Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606946 MIM: 138249 MIM: 613505 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANAPC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ANAPC2 - anaphase promoting complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013366.3 | Intron | NP_037498.1 |
GRIN1 - glutamate ionotropic receptor NMDA type subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC26 - leucine rich repeat containing 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3621 - microRNA 3621 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM210 - transmembrane protein 210 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |