Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PP12613 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PP12613 - uncharacterized LOC100192379 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM155 - transmembrane protein 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317837.1 | Intron | NP_001304766.1 | ||||
NM_001317838.1 | Intron | NP_001304767.1 | ||||
NM_001317839.1 | Intron | NP_001304768.1 | ||||
NM_001317841.1 | Intron | NP_001304770.1 | ||||
NM_001317842.1 | Intron | NP_001304771.1 | ||||
NM_152399.3 | Intron | NP_689612.3 | ||||
XM_005262737.2 | Intron | XP_005262794.1 | ||||
XM_017007725.1 | Intron | XP_016863214.1 | ||||
XM_017007726.1 | Intron | XP_016863215.1 |