Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGGGGGAAGTGGCTTGTCAGCC[C/T]CGGCTCCGGGAAGAGTGAACAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116930 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928847 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101928847 - uncharacterized LOC101928847 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCAM1 - neural cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000615.6 | Intron | NP_000606.3 | ||||
NM_001076682.3 | Intron | NP_001070150.1 | ||||
NM_001242607.1 | Intron | NP_001229536.1 | ||||
NM_001242608.1 | Intron | NP_001229537.1 | ||||
NM_181351.4 | Intron | NP_851996.2 |