Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATTCTAAGGCACCCTTAGCATA[A/G]GGAATTTCAAAATAGACATTTACAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604347 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR186 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR186 - microRNA 186 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZRANB2 - zinc finger RANBP2-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005455.4 | 1803 | UTR 3 | NP_005446.2 | |||
NM_203350.2 | 1803 | UTR 3 | NP_976225.1 |
ZRANB2-AS1 - ZRANB2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |