Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
55 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614123 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMCO1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TMCO1 - transmembrane and coiled-coil domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256164.1 | 3301 | UTR 3 | NP_001243093.1 | |||
NM_001256165.1 | 3301 | UTR 3 | NP_001243094.1 | |||
NM_019026.4 | 3301 | UTR 3 | NP_061899.2 |