Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGTTCCAAGCTGGACGCTATTC[A/G]TTCCTTTGGGCTGGGCTGGCTCAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606822 MIM: 613170 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POMGNT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
POMGNT1 - protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243766.1 | Intron | NP_001230695.1 | ||||
NM_001290129.1 | Intron | NP_001277058.1 | ||||
NM_001290130.1 | Intron | NP_001277059.1 | ||||
NM_017739.3 | Intron | NP_060209.3 | ||||
XM_005271010.1 | Intron | XP_005271067.1 | ||||
XM_006710755.1 | Intron | XP_006710818.1 | ||||
XM_006710756.1 | Intron | XP_006710819.1 | ||||
XM_011541760.2 | Intron | XP_011540062.1 | ||||
XM_017001690.1 | Intron | XP_016857179.1 |
TSPAN1 - tetraspanin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005727.3 | Intron | NP_005718.2 | ||||
XM_011540460.2 | Intron | XP_011538762.1 |
Set Membership: |
HapMap |