Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGTGTATTTGAACAAAAAAAAT[A/G]TGGCTATATTTAGGGTTTTGGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612768 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FNIP2 - folliculin interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323916.1 | Intron | NP_001310845.1 | ||||
NM_020840.2 | Intron | NP_065891.1 | ||||
XM_005263156.1 | Intron | XP_005263213.1 | ||||
XM_005263158.1 | Intron | XP_005263215.1 | ||||
XM_005263160.3 | Intron | XP_005263217.1 | ||||
XM_011532154.2 | Intron | XP_011530456.1 | ||||
XM_017008487.1 | Intron | XP_016863976.1 |