Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
63 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612678 MIM: 611541 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CELF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CELF3 - CUGBP, Elav-like family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172648.3 | Intron | NP_001166119.1 | ||||
NM_001172649.3 | Intron | NP_001166120.1 | ||||
NM_001291106.1 | Intron | NP_001278035.1 | ||||
NM_001291107.1 | Intron | NP_001278036.1 | ||||
NM_007185.6 | Intron | NP_009116.3 | ||||
XM_006711139.2 | Intron | XP_006711202.1 | ||||
XM_006711141.2 | Intron | XP_006711204.1 | ||||
XM_011509111.2 | Intron | XP_011507413.1 | ||||
XM_011509112.2 | Intron | XP_011507414.1 | ||||
XM_011509114.2 | Intron | XP_011507416.1 | ||||
XM_017000145.1 | Intron | XP_016855634.1 | ||||
XM_017000146.1 | Intron | XP_016855635.1 |
SNX27 - sorting nexin family member 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |