Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGACGTTTCATAGGAGCATGAAG[A/G]TAGCTGTTTCCTATTTGTCTGTAGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601020 MIM: 609739 | |||||||||||||||||||||||
Literature Links: |
CD86 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CD86 - CD86 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206924.1 | Intron | NP_001193853.1 | ||||
NM_001206925.1 | Intron | NP_001193854.1 | ||||
NM_006889.4 | Intron | NP_008820.3 | ||||
NM_175862.4 | Intron | NP_787058.4 | ||||
NM_176892.1 | Intron | NP_795711.1 |
ILDR1 - immunoglobulin like domain containing receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |