Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGCAGTGTACCAGAGATTCAGTT[A/G]CTGTGCACCCCACCCTTGCGATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606214 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPTBN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPTBN4 - spectrin beta, non-erythrocytic 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020971.2 | Intron | NP_066022.2 | ||||
NM_025213.2 | Intron | NP_079489.2 | ||||
XM_011527173.2 | Intron | XP_011525475.1 | ||||
XM_017027049.1 | Intron | XP_016882538.1 | ||||
XM_017027050.1 | Intron | XP_016882539.1 | ||||
XM_017027051.1 | Intron | XP_016882540.1 | ||||
XM_017027052.1 | Intron | XP_016882541.1 |