Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGTGAAATGTGTAATATATCC[A/G]TCTTTTTTTATATCGTATTGAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607216 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SDK1 - sidekick cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079653.1 | Intron | NP_001073121.1 | ||||
NM_152744.3 | Intron | NP_689957.3 | ||||
XM_011515188.1 | Intron | XP_011513490.1 | ||||
XM_011515189.1 | Intron | XP_011513491.1 | ||||
XM_011515190.1 | Intron | XP_011513492.1 | ||||
XM_017011837.1 | Intron | XP_016867326.1 | ||||
XM_017011838.1 | Intron | XP_016867327.1 | ||||
XM_017011839.1 | Intron | XP_016867328.1 | ||||
XM_017011840.1 | Intron | XP_016867329.1 |