Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGCACTGAAAAATGGCGGCTCC[A/C]CCTCCCCTAGGCCCTTTCACACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602033 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EML1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EML1 - echinoderm microtubule associated protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008707.1 | Intron | NP_001008707.1 | ||||
NM_004434.2 | Intron | NP_004425.2 | ||||
XM_005267397.1 | Intron | XP_005267454.1 | ||||
XM_005267398.3 | Intron | XP_005267455.1 | ||||
XM_005267399.4 | Intron | XP_005267456.2 | ||||
XM_005267400.4 | Intron | XP_005267457.1 | ||||
XM_011536540.2 | Intron | XP_011534842.1 | ||||
XM_011536541.1 | Intron | XP_011534843.1 | ||||
XM_011536542.2 | Intron | XP_011534844.1 | ||||
XM_017021074.1 | Intron | XP_016876563.1 |