Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATGAAATATTTGTAGTAACTT[C/T]CATAACCCCCTGGGCACCCTCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM149A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM149A - family with sequence similarity 149 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006655.2 | Intron | NP_001006656.1 | ||||
NM_015398.2 | Intron | NP_056213.1 | ||||
XM_005262911.1 | Intron | XP_005262968.1 | ||||
XM_005262912.1 | Intron | XP_005262969.1 | ||||
XM_005262913.1 | Intron | XP_005262970.1 | ||||
XM_011531834.1 | Intron | XP_011530136.1 | ||||
XM_011531835.2 | Intron | XP_011530137.1 | ||||
XM_011531836.1 | Intron | XP_011530138.1 | ||||
XM_017007996.1 | Intron | XP_016863485.1 | ||||
XM_017007997.1 | Intron | XP_016863486.1 | ||||
XM_017007998.1 | Intron | XP_016863487.1 | ||||
XM_017007999.1 | Intron | XP_016863488.1 | ||||
XM_017008000.1 | Intron | XP_016863489.1 | ||||
XM_017008001.1 | Intron | XP_016863490.1 | ||||
XM_017008002.1 | Intron | XP_016863491.1 | ||||
XM_017008003.1 | Intron | XP_016863492.1 | ||||
XM_017008004.1 | Intron | XP_016863493.1 | ||||
XM_017008005.1 | Intron | XP_016863494.1 | ||||
XM_017008006.1 | Intron | XP_016863495.1 | ||||
XM_017008007.1 | Intron | XP_016863496.1 | ||||
XM_017008008.1 | Intron | XP_016863497.1 |