Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603409 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FZD6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FZD6 - frizzled class receptor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164615.1 | 414 | Missense Mutation | ATG,GTG | M,V 33 | NP_001158087.1 | |
NM_001164616.1 | 414 | Missense Mutation | ATG,GTG | M,V 1 | NP_001158088.1 | |
NM_001317796.1 | 414 | UTR 5 | NP_001304725.1 | |||
NM_003506.3 | 414 | Missense Mutation | ATG,GTG | M,V 33 | NP_003497.2 |
Set Membership: |
HapMap |