Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600844 MIM: 174762 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRCOL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRCOL1 - leucine rich colipase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P2RX2 - purinergic receptor P2X 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282164.1 | 505 | Intron | NP_001269093.1 | |||
NM_001282165.1 | 505 | Silent Mutation | ACC,ACT | T,T 156 | NP_001269094.1 | |
NM_012226.4 | 505 | Silent Mutation | ACC,ACT | T,T 84 | NP_036358.2 | |
NM_016318.3 | 505 | Silent Mutation | ACC,ACT | T,T 132 | NP_057402.1 | |
NM_170682.3 | 505 | Silent Mutation | ACC,ACT | T,T 156 | NP_733782.1 | |
NM_170683.3 | 505 | Silent Mutation | ACC,ACT | T,T 156 | NP_733783.1 | |
NM_174872.2 | 505 | Silent Mutation | ACC,ACT | T,T 64 | NP_777361.1 | |
NM_174873.2 | 505 | Silent Mutation | ACC,ACT | T,T 156 | NP_777362.1 | |
XM_005266154.3 | 505 | Silent Mutation | ACC,ACT | T,T 156 | XP_005266211.1 | |
XM_005266155.4 | 505 | Silent Mutation | ACC,ACT | T,T 156 | XP_005266212.1 | |
XM_005266156.4 | 505 | Silent Mutation | ACC,ACT | T,T 156 | XP_005266213.1 | |
XM_011534786.2 | 505 | Silent Mutation | ACC,ACT | T,T 132 | XP_011533088.1 | |
XM_017019035.1 | 505 | Silent Mutation | ACC,ACT | T,T 156 | XP_016874524.1 |
POLE - DNA polymerase epsilon, catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |