Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATCCCACATACTTGTCAGTTGG[A/G]TATTGCAAGAGAGGATGAGTAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162341 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMBR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NMBR - neuromedin B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324307.1 | 1836 | UTR 3 | NP_001311236.1 | |||
NM_001324308.1 | 1836 | UTR 3 | NP_001311237.1 | |||
NM_002511.3 | 1836 | UTR 3 | NP_002502.2 | |||
XM_011535846.1 | 1836 | UTR 3 | XP_011534148.1 | |||
XM_017010901.1 | 1836 | UTR 3 | XP_016866390.1 | |||
XM_017010902.1 | 1836 | UTR 3 | XP_016866391.1 |
Set Membership: |
Validated |