Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCGGGGCTGCAGGGGGAATATT[C/G]CCACCCGGTATCCCTATAGCCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612435 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLCO3A1 - solute carrier organic anion transporter family member 3A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145044.1 | Intron | NP_001138516.1 | ||||
NM_013272.3 | Intron | NP_037404.2 | ||||
XM_005254889.1 | Intron | XP_005254946.1 | ||||
XM_005254891.2 | Intron | XP_005254948.1 | ||||
XM_011521456.2 | Intron | XP_011519758.1 |
Set Membership: |
HapMap |