Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611420 MIM: 602377 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CIZ1 - CDKN1A interacting zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNM1 - dynamin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005336.2 | Intron | NP_001005336.1 | ||||
NM_001288737.1 | Intron | NP_001275666.1 | ||||
NM_001288738.1 | Intron | NP_001275667.1 | ||||
NM_001288739.1 | Intron | NP_001275668.1 | ||||
NM_004408.3 | Intron | NP_004399.2 | ||||
XM_005251763.2 | Intron | XP_005251820.1 | ||||
XM_005251768.2 | Intron | XP_005251825.1 | ||||
XM_005251769.2 | Intron | XP_005251826.1 | ||||
XM_006716992.2 | Intron | XP_006717055.1 | ||||
XM_006716993.1 | Intron | XP_006717056.1 | ||||
XM_011518335.2 | Intron | XP_011516637.1 | ||||
XM_011518336.2 | Intron | XP_011516638.1 | ||||
XM_017014369.1 | Intron | XP_016869858.1 | ||||
XM_017014370.1 | Intron | XP_016869859.1 | ||||
XM_017014371.1 | Intron | XP_016869860.1 | ||||
XM_017014372.1 | Intron | XP_016869861.1 | ||||
XM_017014373.1 | Intron | XP_016869862.1 |
Set Membership: |
HapMap |