Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATGGGATGGGAGTCCAACAAAC[C/G]CAGCCATTGCTCAGACCCCAGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164015 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MATR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MATR3 - matrin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001194954.1 | 169 | UTR 5 | NP_001181883.1 | |||
NM_001194955.1 | 169 | Intron | NP_001181884.1 | |||
NM_001194956.1 | 169 | Intron | NP_001181885.1 | |||
NM_001282278.1 | 169 | UTR 5 | NP_001269207.1 | |||
NM_018834.5 | 169 | Intron | NP_061322.2 | |||
NM_199189.2 | 169 | UTR 5 | NP_954659.1 |
SNHG4 - small nucleolar RNA host gene 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA74A - small nucleolar RNA, H/ACA box 74A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |