Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608342 MIM: 603142 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB5R2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYB5R2 - cytochrome b5 reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302826.1 | 1063 | UTR 3 | NP_001289755.1 | |||
NM_001302827.1 | 1063 | UTR 3 | NP_001289756.1 | |||
NM_016229.4 | 1063 | UTR 3 | NP_057313.2 | |||
XM_005252975.4 | 1063 | UTR 3 | XP_005253032.1 | |||
XM_006718251.2 | 1063 | UTR 3 | XP_006718314.1 | |||
XM_011520184.2 | 1063 | UTR 3 | XP_011518486.1 | |||
XM_011520185.2 | 1063 | UTR 3 | XP_011518487.1 | |||
XM_017017920.1 | 1063 | UTR 3 | XP_016873409.1 | |||
XM_017017921.1 | 1063 | UTR 3 | XP_016873410.1 | |||
XM_017017922.1 | 1063 | UTR 3 | XP_016873411.1 |
Set Membership: |
HapMap |