Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605188 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR85 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR85 - G protein-coupled receptor 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146265.1 | 3521 | UTR 3 | NP_001139737.1 | |||
NM_001146266.1 | 3521 | UTR 3 | NP_001139738.1 | |||
NM_001146267.1 | 3521 | UTR 3 | NP_001139739.1 | |||
NM_018970.6 | 3521 | UTR 3 | NP_061843.3 | |||
XM_005250451.2 | 3521 | Intron | XP_005250508.1 | |||
XM_017012352.1 | 3521 | Intron | XP_016867841.1 |