Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613358 MIM: 611480 MIM: 605208 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH16A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ALDH16A1 - aldehyde dehydrogenase 16 family member A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIH1D1 - PIH1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017916.2 | 771 | UTR 3 | NP_060386.1 | |||
XM_006723254.2 | 771 | UTR 3 | XP_006723317.1 |
SLC17A7 - solute carrier family 17 member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |