Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTTGATGCGGGCTCGTTTCTCA[C/T]GTTCCAGTCTAGAATAAAAAAAGCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604958 MIM: 611852 | |||||||||||||||||||||||
Literature Links: |
ACTL6A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ACTL6A - actin like 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004301.4 | 514 | Intron | NP_004292.1 | |||
NM_177989.3 | 514 | Intron | NP_817126.1 | |||
NM_178042.3 | 514 | Intron | NP_829888.1 |
MRPL47 - mitochondrial ribosomal protein L47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020409.2 | 514 | Missense Mutation | CAT,CGT | H,R 213 | NP_065142.2 | |
NM_177988.1 | 514 | Missense Mutation | CAT,CGT | H,R 103 | NP_817125.1 |