Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTGGACTGCTGGAAGCTAGGGG[A/G]GCCAACTATGGGCTGCATCTAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605547 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSTL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FSTL1 - follistatin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007085.4 | 1987 | UTR 3 | NP_009016.1 |
LOC100130701 - uncharacterized LOC100130701 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR198 - microRNA 198 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |