Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604446 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTER PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTER - phosphotriesterase related | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001484.2 | 66 | UTR 5 | NP_001001484.1 | |||
NM_001261836.1 | 66 | UTR 5 | NP_001248765.1 | |||
NM_001261837.1 | 66 | UTR 5 | NP_001248766.1 | |||
NM_001261838.1 | 66 | UTR 5 | NP_001248767.1 | |||
NM_030664.4 | 66 | UTR 5 | NP_109589.2 | |||
XM_017016927.1 | 66 | Intron | XP_016872416.1 | |||
XM_017016928.1 | 66 | Intron | XP_016872417.1 | |||
XM_017016929.1 | 66 | UTR 5 | XP_016872418.1 | |||
XM_017016930.1 | 66 | Intron | XP_016872419.1 |
Set Membership: |
HapMap |