Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603548 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDS1 - CDP-diacylglycerol synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001263.3 | Intron | NP_001254.2 | ||||
XM_005262687.2 | Intron | XP_005262744.1 | ||||
XM_017007648.1 | Intron | XP_016863137.1 | ||||
XM_017007649.1 | Intron | XP_016863138.1 | ||||
XM_017007650.1 | Intron | XP_016863139.1 | ||||
XM_017007651.1 | Intron | XP_016863140.1 |
Set Membership: |
HapMap |