Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATACTTTGCAATATATAATGAC[A/T]TTTTGCATATTTTATTGTAATCTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603693 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFPM2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFPM2 - zinc finger protein, FOG family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012082.3 | Intron | NP_036214.2 | ||||
XM_011516947.2 | Intron | XP_011515249.1 | ||||
XM_011516948.2 | Intron | XP_011515250.1 |
Set Membership: |
HapMap |