Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCTCACATTTCTTCTTTAGCT[C/G]TTGTTTCATGCTGTGTCCTGTGGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616822 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MON2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MON2 - MON2 homolog, regulator of endosome-to-Golgi trafficking | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278469.1 | Intron | NP_001265398.1 | ||||
NM_001278470.1 | Intron | NP_001265399.1 | ||||
NM_001278471.1 | Intron | NP_001265400.1 | ||||
NM_001278472.1 | Intron | NP_001265401.1 | ||||
NM_015026.2 | Intron | NP_055841.2 | ||||
XM_017019041.1 | Intron | XP_016874530.1 | ||||
XM_017019042.1 | Intron | XP_016874531.1 | ||||
XM_017019043.1 | Intron | XP_016874532.1 | ||||
XM_017019044.1 | Intron | XP_016874533.1 | ||||
XM_017019045.1 | Intron | XP_016874534.1 | ||||
XM_017019046.1 | Intron | XP_016874535.1 |
Set Membership: |
HapMap |