Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATAGCACCACGTGGTCCCGGTC[A/G]TAGCCCCGGCCTCCTGGGGGTGGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114280 MIM: 609151 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIM1L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AIM1L - absent in melanoma 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039775.3 | 5047 | Silent Mutation | TAC,TAT | Y,Y 1637 | NP_001034864.2 | |
XM_005245918.2 | 5047 | Silent Mutation | TAC,TAT | Y,Y 1637 | XP_005245975.1 | |
XM_011541672.1 | 5047 | Silent Mutation | TAC,TAT | Y,Y 1625 | XP_011539974.1 | |
XM_011541673.2 | 5047 | Silent Mutation | TAC,TAT | Y,Y 1694 | XP_011539975.1 |
CD52 - CD52 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBXN11 - UBX domain protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |