Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCAGTGATGAGCATATTGGGCCA[C/T]TTTTTTCCTCAACATGTAAACTCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605142 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
CCT4 - chaperonin containing TCP1 subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256721.1 | 2077 | UTR 3 | NP_001243650.1 | |||
NM_006430.3 | 2077 | UTR 3 | NP_006421.2 |
Set Membership: |
HapMap Validated |