Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCCCAGCTCTTGCTGTCCACCC[C/G]GGCAGTAGGCAGGCAGCCTGGCCAT
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 615647 | |||||||||||||||||||||||||||||
Literature Links: |
TEX19 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
TEX19 - testis expressed 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207459.3 | 374 | UTR 5 | NP_997342.1 | |||
XM_017024638.1 | 374 | UTR 5 | XP_016880127.1 | |||
XM_017024639.1 | 374 | UTR 5 | XP_016880128.1 |