Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606801 MIM: 614632 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAEA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MAEA - macrophage erythroblast attacher | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UVSSA - UV stimulated scaffold protein A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317934.1 | 962 | Silent Mutation | ATC,ATT | I,I 64 | NP_001304863.1 | |
NM_001317935.1 | 962 | Silent Mutation | ATC,ATT | I,I 64 | NP_001304864.1 | |
NM_020894.3 | 962 | Silent Mutation | ATC,ATT | I,I 64 | NP_065945.2 | |
XM_017008490.1 | 962 | Silent Mutation | ATC,ATT | I,I 64 | XP_016863979.1 | |
XM_017008491.1 | 962 | Silent Mutation | ATC,ATT | I,I 64 | XP_016863980.1 | |
XM_017008492.1 | 962 | Silent Mutation | ATC,ATT | I,I 64 | XP_016863981.1 | |
XM_017008493.1 | 962 | Silent Mutation | ATC,ATT | I,I 64 | XP_016863982.1 | |
XM_017008494.1 | 962 | Silent Mutation | ATC,ATT | I,I 64 | XP_016863983.1 | |
XM_017008495.1 | 962 | Intron | XP_016863984.1 | |||
XM_017008496.1 | 962 | Silent Mutation | ATC,ATT | I,I 173 | XP_016863985.1 | |
XM_017008497.1 | 962 | Silent Mutation | ATC,ATT | I,I 173 | XP_016863986.1 | |
XM_017008498.1 | 962 | Silent Mutation | ATC,ATT | I,I 64 | XP_016863987.1 | |
XM_017008499.1 | 962 | Silent Mutation | ATC,ATT | I,I 64 | XP_016863988.1 | |
XM_017008500.1 | 962 | Intron | XP_016863989.1 |