Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGACTGTGGGATTTTATAGACA[G/A]TCTGTTCTCATCAGAGAGATTGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606270 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLR10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TLR10 - toll like receptor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017388.2 | 2559 | Silent Mutation | GAC,GAT | D,D 809 | NP_001017388.1 | |
NM_001195106.1 | 2559 | Silent Mutation | GAC,GAT | D,D 809 | NP_001182035.1 | |
NM_001195107.1 | 2559 | Silent Mutation | GAC,GAT | D,D 809 | NP_001182036.1 | |
NM_001195108.1 | 2559 | Silent Mutation | GAC,GAT | D,D 795 | NP_001182037.1 | |
NM_030956.3 | 2559 | Silent Mutation | GAC,GAT | D,D 809 | NP_112218.2 | |
XM_011513760.2 | 2559 | Silent Mutation | GAC,GAT | D,D 795 | XP_011512062.1 | |
XM_011513761.2 | 2559 | Silent Mutation | GAC,GAT | D,D 809 | XP_011512063.1 | |
XM_011513762.2 | 2559 | Silent Mutation | GAC,GAT | D,D 809 | XP_011512064.1 |