Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614549 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERINC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SERINC2 - serine incorporator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199037.1 | Intron | NP_001185966.1 | ||||
NM_001199038.1 | Intron | NP_001185967.1 | ||||
NM_001199039.1 | Intron | NP_001185968.1 | ||||
NM_018565.3 | Intron | NP_061035.2 | ||||
NM_178865.4 | Intron | NP_849196.2 |
Set Membership: |
HapMap |