Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601288 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
YWHAZ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
YWHAZ - tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135699.1 | Intron | NP_001129171.1 | ||||
NM_001135700.1 | Intron | NP_001129172.1 | ||||
NM_001135701.1 | Intron | NP_001129173.1 | ||||
NM_001135702.1 | Intron | NP_001129174.1 | ||||
NM_003406.3 | Intron | NP_003397.1 | ||||
NM_145690.2 | Intron | NP_663723.1 | ||||
XM_005251061.3 | Intron | XP_005251118.1 | ||||
XM_005251063.3 | Intron | XP_005251120.1 | ||||
XM_011517290.2 | Intron | XP_011515592.1 | ||||
XM_017013810.1 | Intron | XP_016869299.1 | ||||
XM_017013811.1 | Intron | XP_016869300.1 | ||||
XM_017013812.1 | Intron | XP_016869301.1 |
Set Membership: |
HapMap |