Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTTAAAGCTGAAGAGAATAAAC[A/G]TAGTATTTACCCTTGCAGAGGACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615050 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASB5 - ankyrin repeat and SOCS box containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080874.3 | 2926 | UTR 3 | NP_543150.1 | |||
XM_005262759.1 | 2926 | UTR 3 | XP_005262816.1 | |||
XM_011531616.2 | 2926 | UTR 3 | XP_011529918.1 | |||
XM_011531617.2 | 2926 | UTR 3 | XP_011529919.1 |