Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATGGTAAGATAGGCTAACCAG[C/T]TTCTAAGTCCAGACAACTGAGAAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616609 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM65 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM65 - transmembrane protein 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194291.2 | Intron | NP_919267.2 | ||||
XM_011516847.2 | Intron | XP_011515149.1 | ||||
XM_017013104.1 | Intron | XP_016868593.1 | ||||
XM_017013105.1 | Intron | XP_016868594.1 |