Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGGCTCCTCTTCTATCTTCACGC[G/C]CACGCTAGGCCCTGAGCCCAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM87B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM87B - transmembrane protein 87B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032824.2 | 57 | UTR 5 | NP_116213.1 | |||
XM_005263827.2 | 57 | UTR 5 | XP_005263884.1 | |||
XM_006712807.2 | 57 | UTR 5 | XP_006712870.1 | |||
XM_017005121.1 | 57 | Intron | XP_016860610.1 |