Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMCN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HMCN2 - hemicentin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291815.1 | Intron | NP_001278744.1 | ||||
XM_011518465.2 | Intron | XP_011516767.1 | ||||
XM_011518466.2 | Intron | XP_011516768.1 | ||||
XM_011518467.2 | Intron | XP_011516769.1 | ||||
XM_011518468.2 | Intron | XP_011516770.1 | ||||
XM_011518469.2 | Intron | XP_011516771.1 | ||||
XM_011518470.2 | Intron | XP_011516772.1 | ||||
XM_017014585.1 | Intron | XP_016870074.1 | ||||
XM_017014586.1 | Intron | XP_016870075.1 |