Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611458 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GLB1 - galactosidase beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000404.3 | 2427 | UTR 3 | NP_000395.2 | |||
NM_001079811.2 | 2427 | UTR 3 | NP_001073279.1 | |||
NM_001135602.2 | 2427 | UTR 3 | NP_001129074.1 | |||
NM_001317040.1 | 2427 | UTR 3 | NP_001303969.1 |
Set Membership: |
HapMap Validated |