Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
64 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603427 MIM: 604097 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PER3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PER3 - period circadian clock 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UTS2 - urotensin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006786.3 | 775 | Missense Mutation | AAT,AGT | N,S 74 | NP_006777.1 | |
NM_021995.2 | 775 | Missense Mutation | AAT,AGT | N,S 89 | NP_068835.1 | |
XM_011540537.2 | 775 | Missense Mutation | AAT,AGT | N,S 89 | XP_011538839.1 | |
XM_011540538.1 | 775 | Missense Mutation | AAT,AGT | N,S 74 | XP_011538840.1 | |
XM_017000116.1 | 775 | Missense Mutation | AAT,AGT | N,S 89 | XP_016855605.1 | |
XM_017000117.1 | 775 | Missense Mutation | AAT,AGT | N,S 74 | XP_016855606.1 | |
XM_017000118.1 | 775 | Missense Mutation | AAT,AGT | N,S 44 | XP_016855607.1 | |
XM_017000119.1 | 775 | Missense Mutation | AAT,AGT | N,S 89 | XP_016855608.1 |