Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGATAGATCTTGATTCCAAAATGC[A/T]CAATCTCCTCCCGGATCTGACAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603559 MIM: 603696 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTMR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
MTMR4 - myotubularin related protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEPT4 - septin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198713.1 | 739 | Missense Mutation | GAG,GTG | E,V 303 | NP_001185642.1 | |
NM_001256782.1 | 739 | Missense Mutation | GAG,GTG | E,V 326 | NP_001243711.1 | |
NM_001256822.1 | 739 | Missense Mutation | GAG,GTG | E,V 164 | NP_001243751.1 | |
NM_004574.4 | 739 | Missense Mutation | GAG,GTG | E,V 311 | NP_004565.1 | |
NM_080415.3 | 739 | UTR 3 | NP_536340.1 | |||
NM_080416.3 | 739 | Missense Mutation | GAG,GTG | E,V 292 | NP_536341.1 | |
XM_006721949.2 | 739 | Missense Mutation | GAG,GTG | E,V 316 | XP_006722012.1 | |
XM_006721950.3 | 739 | Missense Mutation | GAG,GTG | E,V 217 | XP_006722013.1 | |
XM_006721951.2 | 739 | Missense Mutation | GAG,GTG | E,V 212 | XP_006722014.1 | |
XM_006721952.2 | 739 | Missense Mutation | GAG,GTG | E,V 193 | XP_006722015.1 | |
XM_006721954.2 | 739 | Missense Mutation | GAG,GTG | E,V 164 | XP_006722017.1 | |
XM_006721955.2 | 739 | Missense Mutation | GAG,GTG | E,V 164 | XP_006722018.1 | |
XM_011524911.1 | 739 | Missense Mutation | GAG,GTG | E,V 164 | XP_011523213.1 | |
XM_011524912.1 | 739 | Missense Mutation | GAG,GTG | E,V 164 | XP_011523214.1 |
SEPT4-AS1 - SEPT4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |