Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCCGGCTACTCTTTGCTTATGA[A/G]GGCCCCAGCCTTCCCACCTTCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600749 MIM: 604235 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEBPE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEBPE - CCAAT/enhancer binding protein epsilon | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A8 - solute carrier family 7 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267036.1 | Intron | NP_001253965.1 | ||||
NM_001267037.1 | Intron | NP_001253966.1 | ||||
NM_012244.3 | Intron | NP_036376.2 | ||||
NM_182728.2 | Intron | NP_877392.1 |
Set Membership: |
HapMap |