Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606481 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5J PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
INPP5J - inositol polyphosphate-5-phosphatase J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002837.2 | Intron | NP_001002837.1 | ||||
NM_001284285.1 | Intron | NP_001271214.1 | ||||
NM_001284286.1 | Intron | NP_001271215.1 | ||||
NM_001284287.1 | Intron | NP_001271216.1 | ||||
NM_001284288.1 | Intron | NP_001271217.1 | ||||
NM_001284289.1 | Intron | NP_001271218.1 | ||||
XM_011530143.1 | Intron | XP_011528445.1 | ||||
XM_017028771.1 | Intron | XP_016884260.1 | ||||
XM_017028772.1 | Intron | XP_016884261.1 | ||||
XM_017028773.1 | Intron | XP_016884262.1 |
Set Membership: |
Validated |