Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCTGGGGGCATGATGACGCTGA[A/G]GCAACATGAGAGGGGACGTCTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP92 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFP92 - ZFP92 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136273.1 | 5549 | Intron | NP_001129745.1 | |||
XM_005274652.3 | 5549 | UTR 3 | XP_005274709.1 | |||
XM_011531115.2 | 5549 | UTR 3 | XP_011529417.1 | |||
XM_017029281.1 | 5549 | UTR 3 | XP_016884770.1 |