Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614603 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DDHD1 - DDHD domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160147.1 | 10528 | UTR 3 | NP_001153619.1 | |||
NM_001160148.1 | 10528 | UTR 3 | NP_001153620.1 | |||
NM_030637.2 | 10528 | UTR 3 | NP_085140.2 | |||
XM_005268102.2 | 10528 | Intron | XP_005268159.1 | |||
XM_005268103.2 | 10528 | Intron | XP_005268160.1 | |||
XM_005268105.2 | 10528 | Intron | XP_005268162.1 | |||
XM_011537188.2 | 10528 | UTR 3 | XP_011535490.1 | |||
XM_011537189.2 | 10528 | Intron | XP_011535491.1 | |||
XM_017021668.1 | 10528 | Intron | XP_016877157.1 | |||
XM_017021669.1 | 10528 | Intron | XP_016877158.1 |
Set Membership: |
HapMap Validated |