Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602704 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MDM4 - MDM4, p53 regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204171.1 | 92 | Intron | NP_001191100.1 | |||
NM_001204172.1 | 92 | Intron | NP_001191101.1 | |||
NM_001278516.1 | 92 | Intron | NP_001265445.1 | |||
NM_001278517.1 | 92 | Intron | NP_001265446.1 | |||
NM_001278518.1 | 92 | Intron | NP_001265447.1 | |||
NM_001278519.1 | 92 | Intron | NP_001265448.1 | |||
NM_002393.4 | 92 | Intron | NP_002384.2 | |||
XM_017001311.1 | 92 | UTR 5 | XP_016856800.1 | |||
XM_017001312.1 | 92 | UTR 5 | XP_016856801.1 | |||
XM_017001313.1 | 92 | UTR 5 | XP_016856802.1 |
Set Membership: |
HapMap JSNP Validated |