Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATAAACTGCTGTGTGTCAACTC[A/C]AAATAAAATCCTAAGCCCATCACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604576 MIM: 610865 MIM: 612268 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C14orf1 - chromosome 14 open reading frame 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007176.3 | Intron | NP_009107.1 |
FLVCR2 - feline leukemia virus subgroup C cellular receptor family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTLL5 - tubulin tyrosine ligase like 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |