Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164860 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MET PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MET - MET proto-oncogene, receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000245.3 | Intron | NP_000236.2 | ||||
NM_001127500.2 | Intron | NP_001120972.1 | ||||
NM_001324401.1 | Intron | NP_001311330.1 | ||||
NM_001324402.1 | Intron | NP_001311331.1 | ||||
XM_006715990.2 | Intron | XP_006716053.1 | ||||
XM_011516223.1 | Intron | XP_011514525.1 |
Set Membership: |
HapMap Validated |