Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCCCTTAGAATTTGCTTTTAG[A/C]AATTAACTCAGGTGTTTAGAACATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300515 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
FANCB - Fanconi anemia complementation group B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOSPD2 - motile sperm domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177475.1 | Intron | NP_001170946.1 | ||||
NM_152581.3 | Intron | NP_689794.1 | ||||
XM_005274451.1 | Intron | XP_005274508.1 |